Assay Details
Gene Symbol: CRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198007.1 | NP_001184936.1 | 11 - 12 | 1247 | 71 | |
NM_100718.5 | NP_563816.1 | 12 - 13 | 1387 | 71 | ||
NM_202064.1 | NP_973793.1 | 12 - 13 | 1093 | 71 | ||
GenBank mRNA | AY056320.1 | - | 12 - 13 | 1255 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
