Assay Details
Gene Symbol: OTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331913.1 | NP_001322371.1 | 11 - 12 | 1948 | 72 | |
NM_100932.4 | NP_172527.2 | 12 - 13 | 1568 | 72 | ||
NM_202073.4 | NP_973802.1 | 12 - 13 | 1752 | 72 | ||
GenBank mRNA | AY092981.1 | - | 12 - 13 | 1547 | 72 | |
AY128798.1 | - | 12 - 13 | 1502 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
