Assay Details
Gene Symbol: OTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331913.1 | NP_001322371.1 | 4 - 5 | 1291 | 82 | |
NM_100932.4 | NP_172527.2 | 5 - 6 | 911 | 82 | ||
NM_202073.4 | NP_973802.1 | 5 - 6 | 1095 | 82 | ||
GenBank mRNA | AY092981.1 | - | 5 - 6 | 890 | 82 | |
AY128798.1 | - | 5 - 6 | 845 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
