Assay Details
Gene Symbol: TIF3H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100960.3 | NP_563880.1 | 8 - 9 | 950 | 68 | |
NM_202079.1 | NP_973808.1 | 7 - 8 | 836 | 68 | ||
GenBank mRNA | AY054641.1 | - | 8 - 9 | 784 | 68 | |
AY081546.1 | - | 7 - 8 | 633 | 68 | ||
AY087338.1 | - | 8 - 9 | 852 | 68 | ||
BT000765.1 | - | 8 - 9 | 729 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
