Assay Details
Gene Symbol: TIF3H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100960.3 | NP_563880.1 | 9 - 10 | 1050 | 114 | |
NM_202079.1 | NP_973808.1 | 8 - 9 | 936 | 114 | ||
GenBank mRNA | AY054641.1 | - | 9 - 10 | 884 | 114 | |
AY081546.1 | - | 8 - 9 | 733 | 114 | ||
AY087338.1 | - | 9 - 10 | 952 | 114 | ||
BT000765.1 | - | 9 - 10 | 829 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
