Assay Details
| Gene Symbol: MAP1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101198.4 | NP_172785.1 | 9 - 10 | 1074 | 84 | |
| NM_202086.1 | NP_973815.1 | 8 - 9 | 1110 | 84 | ||
| GenBank mRNA | BT000449.1 | - | 9 - 10 | 1007 | 84 | |
| BT001213.1 | - | 9 - 10 | 964 | 84 | ||
More Information
Set Membership: |
|

MAP1C
Genomic Map