Assay Details
Gene Symbol: AAPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332070.1 | NP_001323307.1 | 13 - 14 | 858 | 86 | |
NM_101226.5 | NP_172813.1 | 15 - 16 | 1227 | 86 | ||
NM_202088.2 | NP_973817.1 | 14 - 15 | 1124 | 86 | ||
GenBank mRNA | AF091843.1 | - | 15 - 16 | 1064 | 86 | |
AY080725.1 | - | 15 - 16 | 1101 | 86 | ||
AY114062.1 | - | 14 - 15 | 965 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
