Assay Details
| Gene Symbol: UBC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101307.5 | NP_563951.1 | 5 - 6 | 705 | 77 | |
| NM_202096.2 | NP_973825.1 | 5 - 6 | 422 | 77 | ||
| GenBank mRNA | AF332451.1 | - | 4 - 5 | 330 | 77 | |
| AK226391.1 | - | 5 - 6 | 552 | 77 | ||
| AY070074.1 | - | 5 - 6 | 503 | 77 | ||
| AY085783.1 | - | 5 - 6 | 551 | 77 | ||
| AY091330.1 | - | 4 - 5 | 330 | 77 | ||
| DQ027016.1 | - | 4 - 5 | 330 | 77 | ||
| M62721.1 | - | 5 - 6 | 429 | 77 | ||
More Information
Set Membership: |
|

UBC1
Genomic Map