Assay Details
Gene Symbol: CLPS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337539.1 | NP_001319471.1 | 4 - 5 | 661 | 82 | |
NM_001337540.1 | NP_001327497.1 | 3 - 4 | 909 | 82 | ||
NM_111340.3 | NP_187119.1 | 4 - 5 | 698 | 82 | ||
GenBank mRNA | AY062866.1 | - | 4 - 5 | 656 | 82 | |
AY128751.1 | - | 3 - 4 | 390 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
