Assay Details
Gene Symbol: SWP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111346.4 | NP_187125.1 | 3 - 4 | 1570 | 101 | |
GenBank mRNA | AF466359.1 | - | 3 - 4 | 1632 | 101 | |
AJ511271.1 | - | 3 - 4 | 1632 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
