Assay Details
Gene Symbol: TIM23-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111352.1 | NP_187131.1 | 1 | 510 | 74 | |
GenBank mRNA | AY463974.1 | - | 1 | 510 | 74 | |
BT025747.1 | - | 1 | 510 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
