Assay Details
| Gene Symbol: AT3G05060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111378.4 | NP_187157.1 | 3 - 4 | 323 | 110 | |
| GenBank mRNA | AF302491.1 | - | 2 - 3 | 126 | 110 | |
| AF412080.1 | - | 3 - 4 | 211 | 110 | ||
More Information
Set Membership: |
|

AT3G05060
Genomic Map