Assay Details
| Gene Symbol: AT3G05060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111378.4 | NP_187157.1 | 6 - 7 | 859 | 75 | |
| GenBank mRNA | AF302491.1 | - | 5 - 6 | 662 | 75 | |
| AF412080.1 | - | 6 - 7 | 747 | 75 | ||
More Information
Set Membership: |
|

AT3G05060
Genomic Map