Assay Details
| Gene Symbol: PLDP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111436.6 | NP_187214.2 | 11 - 12 | 2328 | 95 | |
| GenBank mRNA | AM182458.1 | - | 11 - 12 | 2254 | 95 | |
| AY305003.1 | - | 11 - 12 | 2201 | 95 | ||
More Information
Set Membership: |
|

PLDP2
Genomic Map