Assay Details
| Gene Symbol: PLDP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111436.6 | NP_187214.2 | 13 - 14 | 2771 | 128 | |
| GenBank mRNA | AM182458.1 | - | 13 - 14 | 2697 | 128 | |
| AY305003.1 | - | 13 - 14 | 2644 | 128 | ||
More Information
Set Membership: |
|

PLDP2
Genomic Map