Assay Details
| Gene Symbol: AIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111454.3 | NP_566260.1 | 1 | 946 | 61 | |
| GenBank mRNA | AK176853.1 | - | 1 | 791 | 61 | |
| AY086128.1 | - | 1 | 756 | 61 | ||
More Information
Set Membership: |
|

AIF1
Genomic Map