Assay Details
| Gene Symbol: AT3G06390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111514.3 | NP_187290.1 | 1 - 2 | 403 | 60 | |
| GenBank mRNA | BT003953.1 | - | 1 - 2 | 329 | 60 | |
| BT005725.1 | - | 1 - 2 | 257 | 60 | ||
More Information
Set Membership: |
|

AT3G06390
Genomic Map