Assay Details
| Gene Symbol: CHR11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202897.1 | NP_001189826.1 | 21 - 22 | 2780 | 111 | |
| NM_001202898.2 | NP_001189827.1 | 21 - 22 | 2843 | 111 | ||
| NM_111515.5 | NP_187291.2 | 21 - 22 | 2840 | 111 | ||
More Information
Set Membership: |
|

CHR11
Genomic Map