Assay Details
Gene Symbol: SFR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125119.2 | NP_001118591.1 | 1 - 2 | 291 | 93 | |
NM_111527.3 | NP_566285.1 | 1 - 2 | 374 | 93 | ||
GenBank mRNA | AJ491320.1 | - | 1 - 2 | 290 | 93 | |
AY059889.1 | - | 1 - 2 | 215 | 93 | ||
BT002100.1 | - | 1 - 2 | 170 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
