Assay Details
Gene Symbol: GALK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111534.4 | NP_187310.1 | 11 - 12 | 1514 | 78 | |
GenBank mRNA | AY136356.1 | - | 11 - 12 | 1393 | 78 | |
BT010374.1 | - | 11 - 12 | 1234 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
