Assay Details
Gene Symbol: GALK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111534.4 | NP_187310.1 | 3 - 4 | 664 | 119 | |
GenBank mRNA | AY136356.1 | - | 3 - 4 | 543 | 119 | |
BT010374.1 | - | 3 - 4 | 384 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
