Assay Details
| Gene Symbol: GALK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111534.4 | NP_187310.1 | 4 - 5 | 760 | 68 | |
| GenBank mRNA | AY136356.1 | - | 4 - 5 | 639 | 68 | |
| BT010374.1 | - | 4 - 5 | 480 | 68 | ||
More Information
Set Membership: |
|

GALK
Genomic Map