Assay Details
Gene Symbol: IMPA-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111552.3 | NP_187328.1 | 10 - 11 | 1841 | 92 | |
GenBank mRNA | AY088732.1 | - | 10 - 11 | 1843 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
