Assay Details
| Gene Symbol: ERMO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111590.4 | NP_187366.2 | 8 - 9 | 2729 | 63 | |
| GenBank mRNA | AY136376.1 | - | 8 - 9 | 2608 | 63 | |
| BT000173.1 | - | 7 - 8 | 2088 | 63 | ||
More Information
Set Membership: |
|

ERMO2
Genomic Map