Assay Details
| Gene Symbol: ERMO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111590.4 | NP_187366.2 | 9 - 10 | 2881 | 78 | |
| GenBank mRNA | AY136376.1 | - | 9 - 10 | 2760 | 78 | |
| BT000173.1 | - | 8 - 9 | 2240 | 78 | ||
More Information
Set Membership: |
|

ERMO2
Genomic Map