Assay Details
| Gene Symbol: CSLC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337702.1 | NP_001326755.1 | 3 - 4 | 1111 | 79 | |
| NM_111612.4 | NP_187389.1 | 2 - 3 | 1243 | 79 | ||
| GenBank mRNA | AY070454.1 | - | 2 - 3 | 1100 | 79 | |
| AY142677.1 | - | 2 - 3 | 1017 | 79 | ||
| KJ138836.1 | - | 2 - 3 | 1017 | 79 | ||
More Information
Set Membership: |
|

CSLC6
Genomic Map