Assay Details
| Gene Symbol: AT3G07400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111619.5 | NP_187396.1 | 4 - 5 | 841 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G07400| Gene Symbol: AT3G07400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111619.5 | NP_187396.1 | 4 - 5 | 841 | 70 | |
Set Membership: |
|