Assay Details
| Gene Symbol: IBM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202909.1 | NP_001189838.1 | 11 - 12 | 2646 | 63 | |
| NM_111640.2 | NP_187418.1 | 10 - 11 | 2765 | 63 | ||
More Information
Set Membership: |
|

IBM1
Genomic Map