Assay Details
Gene Symbol: IBM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202909.1 | NP_001189838.1 | 15 - 16 | 3026 | 99 | |
NM_111640.2 | NP_187418.1 | 14 - 15 | 3145 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
