Assay Details
Gene Symbol: IBM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084652.2 | NP_001078121.1 | 4 - 5 | 1054 | 91 | |
NM_001202909.1 | NP_001189838.1 | 4 - 5 | 869 | 91 | ||
NM_111640.2 | NP_187418.1 | 4 - 5 | 1054 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
