Assay Details
Gene Symbol: IBM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202909.1 | NP_001189838.1 | 9 - 10 | 2487 | 92 | |
NM_111640.2 | NP_187418.1 | 8 - 9 | 2606 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
