Assay Details
| Gene Symbol: ADA2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084653.2 | NP_001078122.2 | 5 - 6 | 554 | 96 | |
| NM_001202911.1 | NP_001189840.1 | 7 - 8 | 670 | 96 | ||
| NM_111653.2 | NP_566317.1 | 7 - 8 | 649 | 96 | ||
| NM_202522.1 | NP_974251.1 | 7 - 8 | 735 | 96 | ||
| GenBank mRNA | AF338769.1 | - | 7 - 8 | 625 | 96 | |
| AF360246.1 | - | 7 - 8 | 649 | 96 | ||
| AY040045.1 | - | 7 - 8 | 616 | 96 | ||
More Information
Set Membership: |
|

ADA2A
Genomic Map