Assay Details
| Gene Symbol: QRT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111676.4 | NP_187454.2 | 7 - 8 | 1069 | 139 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

QRT2| Gene Symbol: QRT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111676.4 | NP_187454.2 | 7 - 8 | 1069 | 139 | |
Set Membership: |
|