Assay Details
| Gene Symbol: CIPK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084973.1 | NP_001078442.1 | 2 - 3 | 352 | 94 | |
| NM_118573.4 | NP_194171.1 | 2 - 3 | 629 | 94 | ||
| GenBank mRNA | AF290193.1 | - | 2 - 3 | 229 | 94 | |
| BT000958.1 | - | 2 - 3 | 229 | 94 | ||
More Information
Set Membership: |
|

CIPK8
Genomic Map