Assay Details
| Gene Symbol: CIPK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084973.1 | NP_001078442.1 | 6 - 7 | 650 | 64 | |
| NM_118573.4 | NP_194171.1 | 6 - 7 | 927 | 64 | ||
| GenBank mRNA | BT000958.1 | - | 6 - 7 | 527 | 64 | |
More Information
Set Membership: |
|

CIPK8
Genomic Map