Assay Details
| Gene Symbol: ZIM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118580.4 | NP_194178.1 | 6 - 7 | 1044 | 159 | |
| NM_179104.3 | NP_849435.1 | 7 - 8 | 926 | 159 | ||
| GenBank mRNA | AY086038.1 | - | 6 - 7 | 942 | 159 | |
More Information
Set Membership: |
|

ZIM
Genomic Map