Assay Details
| Gene Symbol: SIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203893.1 | NP_001190822.1 | 2 - 3 | 505 | 65 | |
| NM_001341679.1 | NP_001320056.1 | 3 - 4 | 303 | 65 | ||
| NM_118583.4 | NP_567704.1 | 2 - 3 | 446 | 65 | ||
| GenBank mRNA | AY142488.1 | - | 1 - 2 | 312 | 65 | |
| BT001099.1 | - | 2 - 3 | 225 | 65 | ||
More Information
Set Membership: |
|

SIC
Genomic Map