Assay Details
| Gene Symbol: UBP16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118589.3 | NP_567705.1 | 5 - 6 | 2347 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBP16| Gene Symbol: UBP16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118589.3 | NP_567705.1 | 5 - 6 | 2347 | 64 | |
Set Membership: |
|