Assay Details
Gene Symbol: REN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341680.1 | NP_001320057.1 | 22 - 23 | 2852 | 98 | |
NM_001341681.1 | NP_001329908.1 | 23 - 24 | 2737 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
