Assay Details
Gene Symbol: REN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341680.1 | NP_001320057.1 | 7 - 8 | 1002 | 69 | |
NM_001341681.1 | NP_001329908.1 | 8 - 9 | 887 | 69 | ||
NM_001341682.1 | NP_001329909.1 | 8 - 9 | 659 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
