Assay Details
Gene Symbol: REN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341680.1 | NP_001320057.1 | 8 - 9 | 1115 | 132 | |
NM_001341681.1 | NP_001329908.1 | 9 - 10 | 1000 | 132 | ||
NM_001341682.1 | NP_001329909.1 | 9 - 10 | 772 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
