Assay Details
| Gene Symbol: FC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341698.1 | NP_001329102.1 | 7 - 8 | 825 | 74 | |
| NM_001341700.1 | NP_001329103.1 | 9 - 10 | 1296 | 74 | ||
| NM_001341702.1 | NP_001329105.1 | 8 - 9 | 1976 | 74 | ||
| NM_001341703.1 | NP_001329106.1 | 9 - 10 | 1831 | 74 | ||
| NM_001341704.1 | NP_001329108.1 | 6 - 7 | 1812 | 74 | ||
| NM_001341705.1 | NP_001329109.1 | 9 - 10 | 1359 | 74 | ||
| NM_118607.3 | NP_194205.1 | 10 - 11 | 1259 | 74 | ||
| NM_202881.2 | NP_974610.1 | 9 - 10 | 1069 | 74 | ||
| GenBank mRNA | AK227055.1 | - | 10 - 11 | 1131 | 74 | |
| BT025991.1 | - | 10 - 11 | 1046 | 74 | ||
| D45353.1 | - | 10 - 11 | 1166 | 74 | ||
| U16177.1 | - | 10 - 11 | 1089 | 74 | ||
More Information
Set Membership: |
|

FC2
Genomic Map