Assay Details
| Gene Symbol: FC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341699.1 | NP_001329107.1 | 3 - 4 | 503 | 128 | |
| NM_001341700.1 | NP_001329103.1 | 3 - 4 | 503 | 128 | ||
| NM_001341703.1 | NP_001329106.1 | 2 - 3 | 1109 | 128 | ||
| NM_118607.3 | NP_194205.1 | 3 - 4 | 537 | 128 | ||
| GenBank mRNA | AK227055.1 | - | 3 - 4 | 409 | 128 | |
| BT025991.1 | - | 3 - 4 | 324 | 128 | ||
| D45353.1 | - | 3 - 4 | 444 | 128 | ||
| U16177.1 | - | 3 - 4 | 367 | 128 | ||
More Information
Set Membership: |
|

FC2
Genomic Map