Assay Details
| Gene Symbol: FC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341698.1 | NP_001329102.1 | 5 - 6 | 657 | 67 | |
| NM_001341699.1 | NP_001329107.1 | 8 - 9 | 1057 | 67 | ||
| NM_001341700.1 | NP_001329103.1 | 8 - 9 | 1057 | 67 | ||
| NM_001341701.1 | NP_001329104.1 | 6 - 7 | 1808 | 67 | ||
| NM_001341702.1 | NP_001329105.1 | 6 - 7 | 1808 | 67 | ||
| NM_001341703.1 | NP_001329106.1 | 7 - 8 | 1663 | 67 | ||
| NM_001341704.1 | NP_001329108.1 | 4 - 5 | 1644 | 67 | ||
| NM_001341705.1 | NP_001329109.1 | 7 - 8 | 1191 | 67 | ||
| NM_118607.3 | NP_194205.1 | 8 - 9 | 1091 | 67 | ||
| NM_202881.2 | NP_974610.1 | 7 - 8 | 901 | 67 | ||
| GenBank mRNA | AK227055.1 | - | 8 - 9 | 963 | 67 | |
| BT025991.1 | - | 8 - 9 | 878 | 67 | ||
| D45353.1 | - | 8 - 9 | 998 | 67 | ||
| U16177.1 | - | 8 - 9 | 921 | 67 | ||
More Information
Set Membership: |
|

FC2
Genomic Map