Assay Details
| Gene Symbol: SAE1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118626.3 | NP_567712.1 | 7 - 8 | 795 | 67 | |
| GenBank mRNA | AF510523.1 | - | 7 - 8 | 657 | 67 | |
| AY072375.1 | - | 7 - 8 | 794 | 67 | ||
| AY086568.1 | - | 7 - 8 | 794 | 67 | ||
| BT000094.1 | - | 7 - 8 | 640 | 67 | ||
More Information
Set Membership: |
|

SAE1A
Genomic Map