Assay Details
| Gene Symbol: AT4G25040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118636.3 | NP_194234.1 | 2 - 3 | 450 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G25040| Gene Symbol: AT4G25040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118636.3 | NP_194234.1 | 2 - 3 | 450 | 87 | |
Set Membership: |
|