Assay Details
Gene Symbol: CHLM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125580.1 | NP_001119052.1 | 2 - 3 | 524 | 101 | |
NM_001203903.1 | NP_001190832.1 | 1 - 2 | 726 | 101 | ||
NM_001341721.1 | NP_001328344.1 | 2 - 3 | 650 | 101 | ||
NM_118640.3 | NP_194238.1 | 1 - 2 | 717 | 101 | ||
NM_179107.2 | NP_849438.1 | 1 - 2 | 717 | 101 | ||
NM_179108.3 | NP_849439.1 | 1 - 2 | 717 | 101 | ||
GenBank mRNA | AK316669.1 | - | 1 - 2 | 607 | 101 | |
AK319071.1 | - | 2 - 3 | 519 | 101 | ||
AY034948.1 | - | 1 - 2 | 624 | 101 | ||
AY062991.1 | - | 1 - 2 | 557 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
