Assay Details
| Gene Symbol: SRS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341722.1 | NP_001320059.1 | 11 - 12 | 2023 | 76 | |
| GenBank mRNA | GQ148553.1 | - | 11 - 12 | 1930 | 76 | |
More Information
Set Membership: |
|

SRS2
Genomic Map