Assay Details
| Gene Symbol: SRS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341722.1 | NP_001320059.1 | 18 - 19 | 2726 | 142 | |
| GenBank mRNA | GQ148553.1 | - | 18 - 19 | 2633 | 142 | |
More Information
Set Membership: |
|

SRS2
Genomic Map