Assay Details
Gene Symbol: OLEO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118646.2 | NP_194244.1 | 1 - 2 | 424 | 80 | |
GenBank mRNA | BT010189.1 | - | 1 - 2 | 356 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
